Boron nitride ≥99.5% (metals basis)
Supplier: Thermo Scientific Chemicals
044837-KG
44837-KG
AAA44837-KGEA
452.99
USD
CAAAA44837-KG
AAA44837-KG
Boron nitride ≥99.5% (metals basis)
Boron nitride
Formula:
BN MW: 24.82 g/mol Melting Pt: 2527 °C Density: 2.15 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011317 CAS Number: 10043-11-5 EINECS: 233-136-6 Merck Index: 13,01336 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...