Iridium ≥99.9% (metals basis), wire, Diameter 0.075 mm (0.003 in)

44714-BS 44714-BW
AA44714-BWEA 2185.99 USD
AA44714-BW AA44714-BS
Iridium ≥99.9% (metals basis), wire, Diameter 0.075 mm (0.003 in)
Iridium
Formula: Ir
MW: 192.22 g/mol
Boiling Pt: 5300 °C (1013 hPa)
Melting Pt: 2450 °C
Storage Temperature: Ambient
MDL Number: MFCD00011062
CAS Number: 7439-88-5
EINECS: 231-095-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR