2,3-Dimethoxyphenol 98%

Supplier: Thermo Scientific Chemicals
Warning

44060-04 44060.04
AAA44060-04EA 62.69 USD
AAA44060-04 CAAAA44060-04
2,3-Dimethoxyphenol 98%
2,3-Dimethoxyphenol
Formula: C₈H₁₀O₃
MW: 154.17 g/mol
Boiling Pt: 233…234 °C
Density: 1,182
Flash Pt: 108 °C (226 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008366
CAS Number: 5150-42-5
EINECS: 225-922-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR