Calcium fluoride ≥99.95% (metals basis), sputtering target, Diameter 12.7 mm (0.5 in), Thickness 2.5 mm (0.098 in)

Warning

43343-KS 43343-KS
AAA43343-KSEA 1417.47 USD
CAAAA43343-KS AAA43343-KS
Calcium fluoride ≥99.95% (metals basis), sputtering target, Diameter 12.7 mm (0.5 in), Thickness 2.5 mm (0.098 in)
Calcium fluoride

Magnetron sputtering source

Formula: CaF₂
MW: 78.07 g/mol
Boiling Pt: 2500 °C (1013 hPa)
Melting Pt: 1418 °C
Density: 3,18 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010907
CAS Number: 7789-75-5
EINECS: 232-188-7
Merck Index: 13,01669

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR