Titanium nitride ≥99.5% (metals basis), sputtering target, Diameter 76.2 mm (3.0 in), Thickness 3.18 mm (0.125 in)

Synonyms: Titanium nitride

41112-KS 41112-KS
AAA41112-KSEA 1123.37 USD
CAAAA41112-KS AAA41112-KS
Titanium nitride ≥99.5% (metals basis), sputtering target, Diameter 76.2 mm (3.0 in), Thickness 3.18 mm (0.125 in)
Titanium nitride
Formula: NTi
MW: 61.89 g/mol
Melting Pt: 2950 °C
Density: 5.22
Storage Temperature: Ambient
MDL Number: MFCD00049596
CAS Number: 25583-20-4
EINECS: 247-117-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR