Dipraseodymium trisulfate octahydrate ≥99.99% (REO, rare earth oxide basis)

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Praseodymium(III) sulfate octahydratePraseodymium(III) sulphate octahydrate

013180-14 13180-14
AA13180-14EA 107.99 USD
CAAA13180-14 AA13180-14
Dipraseodymium trisulfate octahydrate ≥99.99% (REO, rare earth oxide basis)
Dipraseodymium trisulphate octahydrate
Formula: O₁₂Pr₂S₃·8H₂O
MW: 714.13 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00149832
CAS Number: 13510-41-3
EINECS: 233-622-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR