Sodium cyanide ≥95% ACS

Supplier: Thermo Scientific Chemicals

Danger

12137-A3 12137-22 12137-36
AAA12137-A3EA 369.85 USD
AAA12137-A3 AAA12137-36 AA12137-22
Sodium cyanide ≥95% ACS
Sodium cyanide
Formula: CNNa
MW: 49.01 g/mol
Boiling Pt: ∼1500 °C
Melting Pt: 563 °C
Density: 1.61
Storage Temperature: Ambient
MDL Number: MFCD00003523
CAS Number: 143-33-9
EINECS: 205-599-4
UN: 1689
ADR: 6.1,I
Merck Index: 13,08679

Order Now

Specification Test Results

Form Granular

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR