Potassium cyanide ≥96.0% ACS
Supplier: Thermo Scientific Chemicals
|
Danger
|
012136-18
012136-A1
012136-30
AAA12136-30EA
97.64
USD
AAA12136-30
AAA12136-A1
AAA12136-18
Potassium cyanide ≥96.0% ACS
Potassium cyanide
Formula:
KCN MW: 65.12 g/mol Boiling Pt: 1625 °C Melting Pt: 634 °C Density: 1.52 Storage Temperature: Ambient |
MDL Number:
MFCD00011397 CAS Number: 151-50-8 EINECS: 205-792-3 UN: 1680 ADR: 6.1,I Merck Index: 13,07709 |
Specification Test Results
Form | Granular |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...