Phosphorus, red ≥98.9% (metals basis), amorphous powder -100 mesh

Supplier: Thermo Scientific Chemicals
Danger

010281-22 010281-36 010281-57
AA10281-22EA 105.51 USD
AA10281-22 AA10281-57 AAA10281-36
Phosphorus, red ≥98.9% (metals basis), amorphous powder -100 mesh
Phosphorus
Formula: P
MW: 30.97 g/mol
Density: ∼2.2 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00133771
CAS Number: 7723-14-0
EINECS: 231-768-7
UN: 1338
ADR: 4.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR