Wasp & Hornet Killer Plus™ Insecticides, CRC
Supplier: CRC
125-14010
300087-876CS 271.76 USD
300087-876
Wasp & Hornet Killer Plus™ Insecticides, CRC
Insecticides
The only wasp and hornet killer in the industry with CRC's patented M.S.D.[L.] , a current MSDS printed on the reverse side of the product label.
  • Will not damage plastic connectors and other insulating materials
  • CRC trigger provides a dense, accurate spray that will reach over 20 feet to drench harmful insects and their nests
VOC 0.1 lb/gal, 15.9 g/L, 2%