You Searched For: Isobutyl propionate
1  results were found
SearchResultCount:"1"
Sort Results
List View Easy View
Rate These Search Results
Supplier: TCI America
Description: CAS Number: 540-42-1 MDL Number: MFCD00009307 Molecular Formula: C7H14O2 Molecular Weight: 130.19 Purity/Analysis Method: 98.0% (GC) Form: Cl...