VACUUBRAND® DCP3000 Mercury Free Vacuum Gauges, BrandTech
Supplier: BrandTech
683170 683190
89178-194EA 3419.43 USD
89178-194 89178-192
VACUUBRAND® DCP3000 Mercury Free Vacuum Gauges, BrandTech
The VACUUBRAND® DCP3000 is a versatile gauge system that allows the use of up to four transducer sets for measurement in the rough and fine vacuum ranges
  • Mercury-free
  • Digital display for accurate readings
  • Quasi-analog display for trend indication
  • Corrosion resistant transducers
  • RS232 port
Choose from two base units: The DCP3000 + VSK3000 is supplied with a capacitive transducer for accurate, gas-independent readings from atmosphere to 0.1 mbar/torr/hPa. The DCP3000 + VSP3000 is supplied with a Pirani thermal conductivity transducer for measurement from atmosphere to 10-3 mbar/Torr/hPa. Additional capacitive or Pirani transducers (sold separately) can be attached to monitor vacuum levels in different applications, or paired for maxium accuracy throughout the gauge range. All transducers are mercury-free. An optional vent valve can also be added.
The DCP3000 display also features a RS232 port for process monitoring and validation.
A digital display allows for accurate readings, while an analog display provides trend indicatgion. Gauge can display in your choice of millibar, Torr or hectopascal.
Ordering information: DCP3000 Gauges are supplied in recyclable packaging with selected transducer, stand and universal (100-230V, 50/60Hz) power supply with US, UK, CEE and Australian plugs.