Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Accessories

Supplier: BECKMAN COULTER
870329




 

ORDER

Expand All / Collapse All
Description Tube Capacity Tube Length Tube Diameter Supplier No. VWR Catalog Number Unit Price Quantity
Round Tube Adapter 15 mL 120 mm 17 mm 870329 BK870329 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

ACCESSORIES

Expand All / Collapse All

Accessories
Description Capacity Tube Capacity Tube Length Tube Diameter Maximum RCF Maximum Speed Supplier No. VWR Catalog Number Unit Price Quantity
Adapter for BD Falcon™ Tubes 9 Tubes 15 ml 120 mm - - 17 mm 368464 BK368464 Pack of 2 Retrieving
Adapter for BD Falcon™ Tubes 4 Tubes 50 ml 120 mm - - 30 mm 368461 BK368461 Pack of 2 Retrieving
25° Conical Fixed Angle Rotor, Model C0650 6 x 50 mL - - 10,400 g (Allegra 64R), 10,304 g (Allegra X-30R), 8346 g (Allegra X-30) 10,000 rpm (Allegra 64R and Allegra X-30R), 8346 rpm (Allegra X-30) - 364670 BK364670 Each Retrieving
25° Conical Fixed Angle Rotor, Model C1015 10 x 15 mL - - 10,400 g (Allegra 64R), 8437 g (Allegra X-30), 10,416 g (Allegra X-30R) 10,000 rpm (Allegra 64R and Allegra X-30R), 9000 (Allegra X-30) - 364680 BK364680 Each Retrieving
25° Fixed Angle Rotor, Model F0685 6 x 85 mL - - 26,320 g (Allegra 64R), 6953 g (Allegra X-30), 10,864 g (Allegra X-30R) 15,500 rpm (Allegra 64R), 8000 rpm (Allegra X-30), 10,000 rpm (Allegra X-30R) - 364650 BK364650 Each Retrieving
25° Fixed Angle Rotor, Model F0850 8 x 50 mL - - 29,220 g (Allegra 64R), 8528 g (Allegra X-30), 13,682 g (Allegra X-30R) 16,500 rpm (Allegra 64R), 9000 rpm (Allegra X-30), 11,400 rpm (Allegra X-30R) - 364640 BK364640 Each Retrieving
25° Fixed Angle Rotor, Model F1010 10 x 10 mL - - 57,440 g (Allegra 64R), 17,896 g (Allegra X-30), 27,579 g (Allegra X-30R) 26,000 rpm (Allegra 64R), 14,500 rpm (Allegra X-30), 18,000 rpm (Allegra X-30R) - 361221 BK361221 Each Retrieving
30° Fixed Angle Rotor, Model F0630 6 x 30 mL - - 59,860 g (Allegra 64R),18,367 g (Allegra X-30), 28,305 g (Allegra X-30R) 26,200 rpm (Allegra 64R), 14,500 (Allegra X-30), 18,000 rpm (Allegra X-30R) - 361231 BK361231 Each Retrieving
45° Fixed Angle Rotor, Model F2402H 24 x 1.5–2.0 mL - - 61,970 g (Allegra 64R), 19,280 g (Allegra X-30), 28,672 g (Allegra X-30R) 26,000 rpm (Allegra 64R), 14,500 rpm (Allegra X-30), 18,000 rpm (Allegra X-30R) - 361171 BK361171 Each Retrieving
Adapter for BD Vacutainer®/Hemolyse/RIA Tubes 16 Tubes 5 mL 75 mm - - 13 mm 368467 BK368467 Pack of 2 Retrieving
Adapter for Bottles with Screw Caps 1 Tube 180 or 200 mL 113 mm - - 56.5 mm 368457 BK368457 Pack of 2 Retrieving
Adapter for Bottles with Screw Caps 1 Tube 125 mL 99 mm - - 50.5 mm 368458 BK368458 Pack of 2 Retrieving
Adapter for Bottles with Screw Caps 1 Tube 85 or 94 mL 112 mm - - 38 mm 392261 BK392261 Each Retrieving
Adapter for Conical-Bottom Tubes 5 Tubes 15 mL 120 mm - - 17 mm 392257 BK392257 Each Retrieving
Adapter for Conical-Bottom Tubes 1 Tube 50 mL 120 mm - - 28.5–29 mm 392268 BK392268 Each Retrieving
Adapter for Flat-Bottom Bottles with Screw Caps 1 Tube 250 mL 125 mm - - 61.8 mm 392256 BK392256 Each Retrieving
Adapter for Glass Bottles with Screw Caps 1 Tube 80 or 100 mL 100 mm - - 44 mm 368459 BK368459 Pack of 2 Retrieving
Adapter for Glass Tubes 5 Tubes 25 mL 100 mm - - 24 mm 368463 BK368463 Pack of 2 Retrieving
Adapter for Glass Tubes 16 Tubes 7 mL 100 mm - - 13 mm 368469 BK368469 Pack of 2 Retrieving
Adapter for Glass Tubes 3 Tubes 50 mL 100 mm - - 34 mm 392265 BK392265 Each Retrieving
Adapter for Microcentrifuge Tubes 16 Tubes 1.5–2.2 mL 38 mm - - 11 mm 368470 BK368470 Pack of 2 Retrieving
Adapter for PCR Tubes 24 Tubes 0.5–0.7 mL 19 mm - - 7 mm 368471 BK368471 Pack of 2 Retrieving
Adapter for Round-Bottom Tubes 12 Tubes 10–15 mL 110 mm - - 17 mm 368465 BK368465 Pack of 2 Retrieving
Adapter for Round-Bottom Tubes 4 Tubes 50 mL 100 mm - - 28.5 mm 368477 BK368477 Pack of 2 Retrieving
Adapter for Tubes with Screw Caps 5 Tubes 30 mL 100 mm - - 26 mm 368462 BK368462 Pack of 2 Retrieving
Adapter for Tubes with Screw Caps 12 Tubes 10 mL 81.1 mm - - 16.1 mm 368468 BK368468 Pack of 2 Retrieving
Bucket Covers, Set of 4, for SX4250 and SX4400 Rotors - - - - - - 392266 BK392266 Each Retrieving
One-Place Adapter for 50 mL Tube - - - - - - 347539 BK347539 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty