Ion Selective Electrodes, Hach
Supplier: Hach
E34M002 LZW9655970002 LZW9650970002 E41M016 LZW9663970002 E41M010 E41M005 LZW9662C.97.002 LZW9652970002 E41M003 LZW9662970002 LZW9662C.97.002
89174-230EA 802.48 USD
89174-230 89174-232 89174-352 89174-330 89174-234 89174-344 CA89501-156 89174-324 89174-236 89174-334 89174-238 89501-156
Ion Selective Electrodes, Hach
Electrodes ISE Electrodes
Hach offers a wide range of electrodes for every application and budget
Our wide selection of metal electrodes, combined metal electrodes, and ion selective electrodes will have the right probe for your application.
Certifications: .CE.WEEE.
Ordering information: The sensION+ electrodes require a reference electrode for use. Many ISE electrodes require standards, filling solutions, ISA, etc. for electrode use.