Lactose

Supplier: USP

Synonyms: Milk sugarD-LactoseAnhydrous Lactose(2R,3R,4R,5R)-2,3,5,6-tetrahydroxy-4-(((2S,3R,4S,5R,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)hexanal

1356676
101095-296EA 496.92 USD
101095-296
Lactose
Lactose
Formula: C₁₂H₂₂O₁₁
MW: 342.3 g/mol
Boiling Pt: 592 °C (1013 hPa)
Melting Pt: 222.8 °C
Storage Temperature: Ambient
MDL Number: MFCD00151251
CAS Number: 63-42-3
EINECS: 200-559-2
Merck Index: 12,05356

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR