Isopropyl palmitate

Supplier: USP

Synonyms: Hexadecylic acid isopropyl esterIsopropyl hexadecanoatePalmitic acid isopropyl esterHexadecanoic acid isopropyl ester

1350603
101095-268EA 453.85 USD
101095-268
Isopropyl palmitate
Isopropyl palmitate
Formula: C₁₉H₃₈O₂
MW: 298.51 g/mol
Boiling Pt: 160 °C (2 mmHg)
Melting Pt: 13,5 °C
Density: 0,855 g/cm³ (20 °C)
MDL Number: MFCD00008993
CAS Number: 142-91-6
EINECS: 205-571-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR