Hydroxyurea

Supplier: USP

Synonyms: N-(Aminocarbonyl)hydroxylamineHydroxycarbamide

1332000
101095-178EA 420 USD
101095-178
Hydroxyurea
Hydroxyurea
Formula: CH₄N₂O₂
MW: 76.05 g/mol
Boiling Pt: 296 °C (1013 hPa)
Melting Pt: 142…144 °C
Storage Temperature: Ambient
MDL Number: MFCD00007943
CAS Number: 127-07-1
EINECS: 204-821-7
Merck Index: 13,04874

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR