Guaifenesin

Supplier: USP

Synonyms: 3-(2-Methoxyphenoxy)-1,2-propanediol3-(2-Methoxyphenoxy)propane-1,2-diolGuaiacol glyceryl ether

1301007
101095-104EA 420 USD
101095-104
Guaifenesin
Guaifenesin
Formula: C₁₀H₁₄O₄
MW: 198.22 g/mol
MDL Number: MFCD00016873
CAS Number: 93-14-1
EINECS: 202-222-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR