Fluorescein

Supplier: USP

Synonyms: 2-(6-Hydroxy-3-oxo-(3H)-xanthen-9-yl)benzoic acid3',6'-Dihydroxy-3H-spiro[isobenzofuran-1,9'-xanthen]-3-one

1277004
101095-030EA 407.69 USD
101095-030
Fluorescein
Fluorescein
Formula: C₂₀H₁₂O₅
MW: 332.31 g/mol
Melting Pt: 314…316 °C
Storage Temperature: Ambient
MDL Number: MFCD00005050
CAS Number: 2321-07-5
EINECS: 219-031-8
Merck Index: 13,04185
https://us.vwr.com/stibo/web/std.lang.all/57/40/2375740.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR