Dopamine hydrochloride

Supplier: USP

Synonyms: 3,4-Dihydroxyphenethylammonium chloride1-(2-Aminoethyl)-3,4-dihydroxybenzene HCl3-Hydroxytyramine hydrochloride3-Hydroxytyraminium chloride

1225204
101094-874EA 420 USD
101094-874
Dopamine hydrochloride
Dopamine hydrochloride
Formula: C₈H₁₂ClNO₂
MW: 189.64 g/mol
Melting Pt: 246…249 °C
Density: 1,356 g/cm³ (20 °C)
MDL Number: MFCD00012898
CAS Number: 62-31-7
EINECS: 200-527-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR