Dibutyl sebacate

Supplier: USP
Warning

Synonyms: Sebacic acid dibutyl ester1,10-Decanedioic acid dibutyl esterDibutyl decanedioate

1187091
101094-798EA 472.31 USD
101094-798
Dibutyl sebacate
Dibutyl sebacate
Formula: C₁₈H₃₄O₄
MW: 314.47 g/mol
Boiling Pt: 180 °C (1013 hPa)
Melting Pt: –11 °C
Density: 0,94 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00027218
CAS Number: 109-43-3
EINECS: 203-672-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR