4-Chloro-3,5-xylenol

Supplier: USP
Warning

Synonyms: Chloroxylenol4-Chloro-3,5-dimethylphenol2-Chloro-5-hydroxy-3-xylene

1122700
101094-066EA 386.15 USD
101094-066
4-Chloro-3,5-xylenol
4-Chloro-3,5-xylenol
Formula: C₈H₉ClO
MW: 156.61 g/mol
Boiling Pt: 246 °C (1013 hPa)
Melting Pt: 114…116 °C
Density: 1,364 g/cm³ (20 °C)
Flash Pt: 132 °C
MDL Number: MFCD00002324
CAS Number: 88-04-0
EINECS: 201-793-8
Merck Index: 13,02194

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR