Butyl-4-hydroxybenzoate

Supplier: USP

Synonyms: ButylparabenButyl parahydroxybenzoate4-Hydroxybenzoic acid butyl esterButyl 4-hydroxybenzoate

1084000
101094-454EA 472.31 USD
101094-454
Butyl-4-hydroxybenzoate
Butyl 4-hydroxybenzoate
Formula: C₁₁H₁₄O₃
MW: 194.23 g/mol
Boiling Pt: 156…157 °C (3.5 mmHg)
Melting Pt: 73 °C
Density: 1.23 g/cm³ (20 °C)
MDL Number: MFCD00016478
CAS Number: 94-26-8
EINECS: 202-318-7
Merck Index: 13,01583

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR