p-Benzoquinone

Supplier: USP
Danger

Synonyms: Quinone1,4-Benzoquinone

1056504
101094-376EA 429.23 USD
101094-376
p-Benzoquinone
p-Benzoquinone
Formula: C₆H₄O₂
MW: 108.1 g/mol
Boiling Pt: ∼174 °C (1013 hPa)
Melting Pt: 112…114 °C
Density: 1.318 g/cm³ (20 °C)
Flash Pt: 77 °C
Storage Temperature: Ambient
MDL Number: MFCD00001591
CAS Number: 106-51-4
EINECS: 203-405-2
UN: 2587
ADR: 6.1,II
Merck Index: 12,08259
https://us.vwr.com/stibo/web/std.lang.all/69/87/2296987.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR