Benzocaine (ethyl-4-aminobenzoate)

Supplier: USP
Warning

Synonyms: AethoformEthyl 4-aminobenzoate

1054000
101094-370EA 420 USD
101094-370
Benzocaine (ethyl-4-aminobenzoate)
Benzocaine (Ethyl 4-aminobenzoate)
Formula: C₉H₁₁NO₂
MW: 165.19 g/mol
Boiling Pt: 311 °C (1013 hPa)
Melting Pt: 89…90 °C
Flash Pt: 112 °C
Storage Temperature: Ambient
MDL Number: MFCD00007892
CAS Number: 94-09-7
EINECS: 202-303-5
Merck Index: 13,01087

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR