Phenazone
Supplier: USP
|
Warning
|
Synonyms:
Antipyrine
, 2,3-Dimethyl-1-phenyl-3-pyrazolin-5-one
1040005
101094-308EA
407.69
USD
101094-308
Phenazone
Phenazone
Formula:
C₁₁H₁₂N₂O MW: 188.23 g/mol Boiling Pt: 388 °C (1013 hPa) Melting Pt: 152…154 °C Density: 1,25 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00003146 CAS Number: 60-80-0 EINECS: 200-486-6 Merck Index: 12,00757 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...