4-Aminohippuric acid ≥98%

Supplier: USP

Synonyms: p-Aminohippuric acid2-(4-Aminobenzamido)acetic acid

1025351
101094-268EA 384.62 USD
101094-268
4-Aminohippuric acid ≥98%
4-Aminohippuric acid
Formula: H₂NC₆H₄C(O)NHCH₂COOH
MW: 194.19 g/mol
Boiling Pt: 441 °C (1013 hPa)
Melting Pt: 199…202 °C
Density: 1.402 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007890
CAS Number: 61-78-9
EINECS: 200-518-9
Merck Index: 13,00441

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR