2-Amino-5-chlorobenzophenone ≥99%

Supplier: USP

Synonyms: (2-Amino-5-chlorophenyl)(phenyl)methanone1-Amino-4-chloro-2-(phenylmethanoyl)benzene

1022808
101096-774EA 1389.23 USD
101096-774
2-Amino-5-chlorobenzophenone ≥99%
2-Amino-5-chlorobenzophenone
Formula: C₁₃H₁₀ClNO
MW: 231.68 g/mol
Boiling Pt: 424 °C (1013 hPa)
Melting Pt: 96…99 °C
MDL Number: MFCD00007839
CAS Number: 719-59-5
EINECS: 211-949-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR