Aluminium sulfate hydrate

Supplier: USP
Danger

Synonyms: Aluminum(III) sulfate xhydrateAluminum(III) sulphate xhydrate

1017364
101094-248EA 287.69 USD
101094-248
Aluminium sulfate hydrate
Aluminium sulphate hydrate
Formula: Al₂(SO₄)₃·xH₂O
MW: 366,21 g/mol
MDL Number: MFCD00149136
CAS Number: 17927-65-0
EINECS: 233-135-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR