N-Acetyl-L-cysteine

Supplier: USP

Synonyms: Acetylcysteine(R)-2-Acetamido-3-mercaptopropanoic acid(R)-(+)-N-AcetylcysteineAc-Cys-OH

1009005
101094-208EA 420 USD
101094-208
N-Acetyl-L-cysteine
N-Acetyl-L-cysteine
Formula: C₅H₉NO₃S
MW: 163.2 g/mol
Boiling Pt: 371 °C (1013 hPa)
Melting Pt: 106…108 °C
Density: 1.48 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004880
CAS Number: 616-91-1
EINECS: 210-498-3
Merck Index: 13,00090

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR