Apigenin

Supplier: R&D Systems

Synonyms: 5,7-Dihydroxy-2-(4-hydroxyphenyl)-4-benzopyrone4',5,7-Trihydroxyflavone5,7-Dihydroxy-2-(4-hydroxyphenyl)-4H-chromen-4-one4',6'-Dihydroxy-2-(4-hydroxyphenyl)benzo(e)oxacyclohex-2-en-4-on5,7-Dihydroxy-2-(4-hydroxyphenyl)-4H-1-benzopyran-4-one5,7-Dihydroxy-2-(4-hydroxyphenyl)chromen-4-one

1227/50 1227/10
100505-282EA 125.82 USD
100505-282 100507-368
Apigenin
Apigenin
Formula: C₁₅H₁₀O₅
MW: 270.24 g/mol
MDL Number: MFCD00006831
CAS Number: 520-36-5
EINECS: 208-292-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR