You Searched For: Ammonium nitrate


16  results were found

SearchResultCount:"16"

Sort Results

List View Easy View

Rate These Search Results

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Description: Ammonium Nitrate, Crystal, AR® (ACS)
Catalog Number: (EM1.01187.5000)
Supplier: MilliporeSigma
Description: Emplura* Ammonium nitrate, CAS No: 6484-52-2, Chemical Formula: NH4NO3, Molar Mass: 80.04 g/mol, Grade: extra pure, Synonyms: Nitric acid ammonia, Ammonia nitrate, pH value: 4.5 - 7.0, Concentration: </=100%, Storage: 5 deg C to 30 deg C, Pack size: 5 kg

Catalog Number: (EM1.02596.2500)
Supplier: MilliporeSigma
Description: AMMONIUM NITRATE SOLUTI (2.5L)

Supplier: MilliporeSigma
Description: AMMONIUM NITRATE FOR ANALYSIS
Supplier: Thermo Scientific Chemicals
Description: AMMONIUM NITRATE 1 KG

Catalog Number: (RCMMAN5N-50)
Supplier: Ricca Chemical
Description: 5%. Used to support Inductively coupled plasma (ICP) analysis to sharpen the contrast and improve the readability. 50mL.

Supplier: Thermo Scientific Chemicals
Description: Puratronic*, 99.999% (metals basis),UN1942,Crystalline,DANGER: OXIDIZER, irritates skin and eyes,50g,250g,OXIDIZER,CAN EVERY SHIPMENT - NO CHARGE FOR CAN
Catalog Number: (RCRV010132-50N)
Supplier: Ricca Chemical
Description: STANDARD NH4NO3 VERISPEC 50ML AAS IN H2O

SDS


Supplier: Thermo Scientific Chemicals
Description: AMMONIUM NITRATE 1KG

Supplier: Honeywell Research Chemicals
Description: Ammonium nitrate, Purity: >/=98%, Grade: Puriss. P.a, ACS Reagent, Reag. ISO, Reag. Ph. Eur, Cas: 6484-52-2, MF: NH4NO3, Molar Mass: 80.04 g/mol, Synonym: Laboratory Chemical, Container: Poly bottle, Color/Form: Colorless or White

SDS

Supplier: Thermo Scientific Chemicals
Description: 2kg MFCD00011425
Supplier: BeanTown Chemical
Description: Ammonium nitrate, Grade: ACS, Purity: 95% Minimum, CAS Number: 6484-52-2, Molecular Formula: H4N2O3, Size: 2Kg

SDS

Supplier: BeanTown Chemical
Description: Ammonium nitrate, Purity: 99.999%, Trace metal Basis, Cas Number: 6484-52-2, Molecular Formula: H4N2O3, Size: 10G

SDS

Supplier: Thermo Scientific Chemicals
Description: AMMONIUM NITRATE 100GR

Supplier: Strem Chemicals Inc
Description: CAS #: 6484-52-2. Size: 2000g.

Supplier: Rigaku Reagents
Description: Use our stock solutions directly or as a component in solutions you create in your own lab. We prepare our stock solutions using high quality raw mate...

Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at 1-800-932-5000.
1 - 16 of 16
no targeter for Bottom