You Searched For: ortho-Boric acid


56  results were found

SearchResultCount:"56"

Sort Results

List View Easy View

Rate These Search Results

Supplier: VWR International
Description: Meets reagent specifications for testing USP/NF monographs.
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Description: Granular. Lot analysis on label.
Supplier: VWR
Description: Biological buffers are useful for cell culture in vitro, enzyme assays, and some electrophoretic applications at physiological pH.
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Description: Boric Acid, Granular, AR® (ACS)
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Description: Boric Acid, Granular, N.F., Multi-Compendial
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Description: GMP manufactured.
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Description: Boric Acid, Granular, N.F.
Catalog Number: (MK139420)
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Description: GMP manufactured.

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Description: For molecular biology buffers. Lot analysis on label.
Catalog Number: (JT0096-05)
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Description: Boric Acid, Granular, N.F.

Supplier: MilliporeSigma
Description: Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs
Supplier: MilliporeSigma
Description: BORIC ACID FOR ANALYSIS (1KG)
Supplier: MilliporeSigma
Description: BORIC ACID 99.9999 SUPRAPUR 50
Catalog Number: (EM1.00165.9025)
Supplier: MilliporeSigma
Description: BORIC ACID FOR ANALYSIS EMSURE 25KG

SDS


Supplier: MP Biomedicals
Description: Boric acid is used for weatherproofing wood and fireproofing fabrics; as a preservative; manufacturing of cements, crockery, porcelain, enamels, glass...

SDS

Supplier: Thermo Scientific Chemicals
Description: 99.99% (metals basis).+8 Mesh Granular,WARNING: Irritates eyes, lungs, may be harmful if swallowed,500g,2kg.
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at 1-800-932-5000.
1 - 16 of 56
no targeter for Bottom