    Sort by:
  • Search Within Results
(S)-(-)-Epoxystyrene
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
SearchResultCount:"1"
  List View Easy View
Sort by:
Supplier:  Acros Organics
Description:   (S)-Styrene oxide, 94%, CAS No 20780-54-5, Molecular weight 120.15, Molecular formula C8 H8 O, Appearance (Color) Colorless to light yellow, Appearance (Form) Liquid, Synonyms: (S)-(-)-Phenyloxirane, (S)-Phenylethylene oxide, Size: 25G