Zinc oxide ≥99%, powder ACS
Supplier: SPECTRUM CHEMICAL MFG CORP
|
Warning
|
Synonyms:
Zinci oxidum
, Zinc (II) oxide
Zinc Oxide, Powder, Reagent, ACS is a metallic chemical element with the atomic number of 30 and has a variety of uses. High volume usages of zinc oxide are in rubber and ceramic glaze manufacturing but it is also used as a UV absorber in suntan lotions and sunglasses, a corrosion inhibitor in paints and as nutrient supplement.
As an ACS grade quality reagent, its chemical specifications are the de facto standards for chemicals used in many high-purity applications and typically designate the highest quality chemical available for laboratory use. Spectrum Chemical manufactured Reagent ACS grade products meet the toughest regulatory standards for quality and purity.
Formula:
ZnO MW: 81.39 g/mol Melting Pt: 1975 °C Density: 5.61 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011300 CAS Number: 1314-13-2 EINECS: 215-222-5 UN: 3077 ADR: 9,III REACH: 01-2119463881-32 Merck Index: 13,10200 |
Specification Test Results
Assay | 99% Min. |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...