Beeswax, white, cake NF

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: Wax, bees

W1010-1CS12 W1010-1LB
95036-576EA 190 USD
95036-576 95036-574
Beeswax, white, cake NF
Beeswax

All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP). 

Melting Pt: 61…65 °C
Density: 0.95 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00132754
CAS Number: 8012-89-3
EINECS: 232-383-7
Merck Index: 12,01049

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR