DL-Valine ≥98%

Synonyms: (±)-ValineH-DL-Val-OH2-Amino-3-methylbutanoic acid

VA115-1KG VA115-500GM VA115-25GM VA115-100GM
700007-432EA 110.72 USD
700007-432 700007-438 700007-434 700007-436
DL-Valine ≥98%
DL-Valine

DL-Valine, also known as 2-amino-3-methylbutanoic acid, is a branched-chain amino acid.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: C₅H₁₁NO₂
MW: 117.15 g/mol
Boiling Pt: 217 °C (1013 hPa)
Melting Pt: 86 °C
Density: 1.352 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004267
CAS Number: 516-06-3
EINECS: 208-220-0
Merck Index: 12,10046

Order Now

Specification Test Results

Assay 98% Min.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR