L(+)-Valine ≥98.5% USP
Supplier: SPECTRUM CHEMICAL MFG CORP
Synonyms:
(S)-(+)-Valine
, Valine
, H-Val-OH
V1005-500GM
V1005-25GM
V1005-1KG
V1005-100GM
95034-116EA
698
USD
95034-116
95034-114
95034-112
95034-110
L(+)-Valine ≥98.5% USP
L(+)-Valine
All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Formula:
C₅H₁₁NO₂ MW: 117.15 g/mol Boiling Pt: 217 °C (1013 hPa) Melting Pt: 86 °C Density: 1.284 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00064220 CAS Number: 72-18-4 EINECS: 200-773-6 Merck Index: 12,10046 |
Specification Test Results
Assay | 98.5% Min. |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...