TRITON™ X-405 (Polyethylene glycol tert-octylphenyl ether) solution 70% in water ≥70%

Supplier: SPECTRUM CHEMICAL MFG CORP
Danger

TR140-4LT TR140-20LTMT TR140-500ML
700011-190EA 469 USD
700011-190 700011-192 700011-188
TRITON™ X-405 (Polyethylene glycol tert-octylphenyl ether) solution 70% in water ≥70%
TRITON™ X-405 (Polyethylene glycol tert-octylphenyl ether)

Triton(R) X-405, 70 Percent Solution in Water is used as an excellent emulsion stabilizer that provides freeze/thaw and ionic stability.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: C₂₈H₅₀O₈
MW: 514.69 g/mol
Density: 1.096
Storage Temperature: Ambient
MDL Number: MFCD00128254
CAS Number: 9036-19-5

Order Now

Specification Test Results

ACTIVE CONTENT 69-71 %
pH OF A 1% SOLUTION IN 62.5% IPA @ 25°C 7.0-9.0
COLOR (APHA) max 250

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR