4-Tolualdehyde ≥99%

Supplier: SPECTRUM CHEMICAL MFG CORP
Warning

Synonyms: 4-Methylbenzaldehydp-Tolualdehyde4-Tolylaldehydep-Methylbenzaldehyd4-Methylbenzaldehydep-Tolylaldehyde

T2376-100GM T2376-500GM T2376-25GM
700003-806EA 186.97 USD
700003-806 700003-802 700003-804
4-Tolualdehyde ≥99%
4-Tolualdehyde
Formula: C₈H₈O
MW: 120.15 g/mol
Boiling Pt: 205 °C (1013 hPa)
Melting Pt: –6 °C
Density: 1,019
Flash Pt: 72 °C
Storage Temperature: Ambient
MDL Number: MFCD00006954
CAS Number: 104-87-0
EINECS: 203-246-9

Order Now

Specification Test Results

Assay 99% Min.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR