1,3,5-Trimethoxybenzene ≥99%
Supplier: SPECTRUM CHEMICAL MFG CORP
Synonyms:
O,O,O-1,3,5-Trimethylresorcinol
T1351-10GM
T1351-50GM
T1351-250GM
700003-438EA
150.32
USD
700003-438
700003-434
700003-436
1,3,5-Trimethoxybenzene ≥99%
1,3,5-Trimethoxybenzene
1,3,5-Trimethoxybenzene or phloroglucinol trimethyl ether is used as a cosmetic agent in skin conditioning products.
Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.
Formula:
C₉H₁₂O₃ MW: 168.19 g/mol Boiling Pt: 255 °C (1013 hPa) Melting Pt: 52…54 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008385 CAS Number: 621-23-8 EINECS: 210-673-4 |
Specification Test Results
Assay | 99% Min. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...