Melissic acid ≥99%

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: Triacontanoic acid

T1319-100MG
700005-328EA 109 USD
700005-328
Melissic acid ≥99%
Mellissic acid

Triacontanoic Acid is used as a viscosity control agent.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: C₃₀H₆₀O₂
MW: 452.81 g/mol
Melting Pt: 97…99 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00002813
CAS Number: 506-50-3
EINECS: 208-042-3

Order Now

Specification Test Results

Assay 99% Min.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR