Triethylene glycol, purified
Supplier: SPECTRUM CHEMICAL MFG CORP
Synonyms:
2,2'-(Ethane-1,2-diylbis(oxy))diethanol
, 2,2'-(Ethylenedioxy)diethanol
T1118-4LTGL
T1118-200LTB
T1118-20LTMT
T1118-500MLG
700003-560EA
1083
USD
700003-560
700003-558
700003-562
700003-564
Triethylene glycol, purified
Triethylene glycol
Triethylene Glycol, Purified, also known as Triglycol, can be used as a liquid desiccant for natural gas and in air conditioning systems. The Purified Grade describes chemicals of good quality where there are no official standards and is normally limited to inorganic chemicals.
Formula:
C₆H₁₄O₄ MW: 150.17 g/mol Boiling Pt: 285 °C (1013 hPa) Melting Pt: –7 °C Density: 1.125 Storage Temperature: Ambient |
MDL Number:
MFCD00002880 CAS Number: 112-27-6 EINECS: 203-953-2 Merck Index: 12,09802 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...