Gum tragacanth No. 1 Ribbon, ribbon FCC

T1088-125GM T1088-500GM
95036-346EA 304.16 USD
95036-346 95036-348
Gum tragacanth No. 1 Ribbon, ribbon FCC
Gum tragacanth

Tragacanth, No. 1 Ribbon, FCC is used as an emulsifier, thickener, stabilizer, and texturant additive. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage, nutritional supplements and personal care applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Storage Temperature: Ambient
MDL Number: MFCD00131255
CAS Number: 9000-65-1
EINECS: 232-552-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR