Talc, powder USP
Supplier: SPECTRUM CHEMICAL MFG CORP
|
Warning
|
Synonyms:
Soapstone
, Magnesium silicate monohydrate
, Talcum venetum
T1005-500GM
T1005-45KGBL
T1005-2.5KG
T1005-12KG
T1005-500G
95036-180EA
279
USD
CA11027-354
95036-180
89050-528
95036-182
89050-530
Talc, powder USP
Talc
All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Formula:
Mg₃O₁₁Si₄·1H₂O MW: 379.27 g/mol Melting Pt: >1200 °C Density: 2.58…2.83 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00792903 CAS Number: 14807-96-6 EINECS: 238-877-9 Merck Index: 13,09127 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...