Sudan II

Synonyms: 1-((2,4-Dimethylphenyl)diazenyl)naphthalen-2-ol1-(2,4-Dimethylphenylazo)-2-naphtholSolvent Orange 7

SU110-25GM
700010-338EA 111.91 USD
700010-338
Sudan II
Sudan II

Sudan II, a lysochrome diazo dye, is related to azobenzene. It can be used to color nonpolar substances such as fats, oils, waxes, and hydrocarbon products.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: C₁₈H₁₆N₂O
MW: 276.34 g/mol
Melting Pt: 156…158 °C
Storage Temperature: Ambient
MDL Number: MFCD00003896
CAS Number: 3118-97-6
EINECS: 221-490-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR