Silicon dioxide amorphous, crystalline 325 Mesh

Supplier: SPECTRUM CHEMICAL MFG CORP
Danger

Synonyms: Silicon(IV) oxideSilicium (IV) oxideSilicium dioxide

SI108-500GM SI108-45KGBL SI108-2.5KG SI108-12KG
700000-698EA 100 USD
700000-698 700000-692 700000-694 700000-696
Silicon dioxide amorphous, crystalline 325 Mesh
Silicium dioxide

Silicon Dioxide, 325 Mesh, Crystalline is also referred to as silica. Found naturally under the form of quartz and in some living organism, it is one of the most complex and abundant families of materials. Nearly 95% of all silicon dioxide is used in the construction industry in the form of glass and Portland cement among others. It is also used as a powder flow helper in the production of tablets

Formula: SiO₂
MW: 60.08 g/mol
Boiling Pt: 2230 °C (1013 hPa)
Melting Pt: 1427 °C
Density: 2.622…2.645 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011232
CAS Number: 14808-60-7
EINECS: 238-878-4

Order Now

Specification Test Results

Particle size 325 Mesh

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR