Sebacic acid ≥94%, Reagent Grade

Synonyms: Decanedioic acid1,8-Octanedicarboxylic acid1,10-Decanedioic acid

SE105-45KGBL SE105-500GM SE105-100GM
700005-834EA 67.79 USD
700005-834 700005-836 700005-838
Sebacic acid ≥94%, Reagent Grade
Sebacic acid

Sebacic Acid, Reagent, also known as 1,8-Octanedicarboxylic acid, is a dicarboxylic acid and can be used as an intermediate for antiseptics and aromatics. The Reagent grade denotes that this chemical is the highest quality commercially available and that the American Chemical Society has not officially set any specifications for this material. Spectrum Chemical manufactured Reagent grade products meet the toughest regulatory standards for quality and purity.

Formula: C₁₀H₁₈O₄
MW: 202.25 g/mol
Boiling Pt: 374 °C (1013 hPa)
Melting Pt: 127…131 °C
Density: 1.21 g/cm³ (20 °C)
Flash Pt: 221 °C
Storage Temperature: Ambient
MDL Number: MFCD00004440
CAS Number: 111-20-6
EINECS: 203-845-5
REACH: 01-2119519212-52
Merck Index: 12,08558

Order Now

Specification Test Results

Assay 94% Min.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR