Sodium bromate ≥99%, crystals, Reagent Grade

Supplier: SPECTRUM CHEMICAL MFG CORP
Danger

S1190-12KGBL S1190-25GM S1190-125GM S1190-50KGBL S1190-500GM
700001-376EA 2257 USD
700001-376 700001-374 700001-372 700001-370 700001-368
Sodium bromate ≥99%, crystals, Reagent Grade
Sodium bromate

Sodium Bromate, Crystal, Reagent, also known as Bromic acid, is a strong oxidant mainly used as a chemical agent. The Reagent grade denotes that this chemical is the highest quality commercially available and that the American Chemical Society has not officially set any specifications for this material. Spectrum Chemical manufactured Reagent grade products meet the toughest regulatory standards for quality and purity.

Formula: BrNaO₃
MW: 150.89 g/mol
Boiling Pt: 1390 °C (1013 hPa)
Melting Pt: 381 °C
Density: 3.254 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003476
CAS Number: 7789-38-0
EINECS: 232-160-4
UN: 1494
ADR: 5.1,II
Merck Index: 12,08736

Order Now

Specification Test Results

Assay 99% Min.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR