Sodium octadecyl sulfate ≥93%
Supplier: SPECTRUM CHEMICAL MFG CORP
|
Warning
|
Synonyms:
Octadecyl sulphuric acid salt
, Sodium Stearyl sulphate
, Sodium Stearyl sulfate
, Sodium n-octadecylsulphate
, n-Octadecyl sulphate, sodium salt
, Octadecyl sulfuric acid salt
, n-Octadecyl sulfate, sodium salt
, Sodium n-octadecylsulfate
S1166-1GM
700006-710EA
96
USD
700006-710
Sodium octadecyl sulfate ≥93%
Sodium octadecyl sulphate
Formula:
C₁₈H₃₇NaO₄S MW: 372.55 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00007469 CAS Number: 1120-04-3 EINECS: 214-295-0 |
Specification Test Results
Assay | 93% Min. |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...